ENA371177964 Enamine

N-(2-amino-4-chlorophenyl)-3-[(4-methylpentan-2-yl)oxy]propanamide

  • MDL number MFCD09803365
  • Cas Number : 1016745-49-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-105677

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-105677
Availability details for
Quantity
  • Availability
  • Add to Cart