ENA371177968 Enamine

2-(3-methylbutyl)piperidine hydrochloride

  • MDL number MFCD22375370
  • Cas Number : 14162-70-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-107756

Safety Information

msds https://www.enaminestore.com/catalog/EN300-107756
Availability details for
Quantity
  • Availability
  • Add to Cart