ENA371177973 Enamine

3-bromo-2-iodobenzoic acid

  • MDL number MFCD15527220
  • Cas Number : 503821-94-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 139...141
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-107770

Safety Information

msds https://www.enaminestore.com/catalog/EN300-107770
Availability details for
Quantity
  • Availability
  • Add to Cart