ENA371177980 Enamine

4-bromo-3-(3,4-dimethylphenyl)-1H-pyrazole

  • MDL number MFCD14657920
  • Cas Number : 1250379-75-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 116...118
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-107824

Safety Information

msds https://www.enaminestore.com/catalog/EN300-107824
Availability details for
Quantity
  • Availability
  • Add to Cart