ENA371177982 Enamine

3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)thiophene-2-carbonitrile

  • MDL number MFCD12405473
  • Cas Number : 942070-34-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-107826

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-107826
Availability details for
Quantity
  • Availability
  • Add to Cart