ENA371177985 Enamine

1,1-dioxo-1lambda6-benzothiophene-5-carbonitrile

  • MDL number MFCD20259134
  • Cas Number : 1394040-24-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-105848

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-105848
Availability details for
Quantity
  • Availability
  • Add to Cart