ENA371177987 Enamine

4-bromo-3-(4-chlorophenyl)-1H-pyrazole

  • MDL number MFCD11226935
  • Cas Number : 17978-27-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 140...142
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-107832

Safety Information

msds https://www.enaminestore.com/catalog/EN300-107832
Availability details for
Quantity
  • Availability
  • Add to Cart