ENA371177993 Enamine

1-(3-methyl-1,1-dioxo-1$l^{6}-thiolan-3-yl)-1H-pyrazole-4-carbaldehyde

  • MDL number MFCD12768931
  • Cas Number : 1184038-95-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 84...86
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-107841

Safety Information

msds https://www.enaminestore.com/catalog/EN300-107841
Availability details for
Quantity
  • Availability
  • Add to Cart