ENA371177997 Enamine

2-(diethylamino)ethyl 2-phenylbutanoate

  • MDL number MFCD00865223
  • Cas Number : 14007-64-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-107845

Safety Information

msds https://www.enaminestore.com/catalog/EN300-107845
Availability details for
Quantity
  • Availability
  • Add to Cart