ENA371178001 Enamine

N-(but-2-yn-1-yl)cyclopentanamine

  • MDL number MFCD16705559
  • Cas Number : 1247601-37-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-107849

Safety Information

msds https://www.enaminestore.com/catalog/EN300-107849
Availability details for
Quantity
  • Availability
  • Add to Cart