ENA371178005 Enamine

N-(but-3-yn-1-yl)cyclopentanamine

  • MDL number MFCD18841175
  • Cas Number : 1394041-84-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-107854

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-107854
Availability details for
Quantity
  • Availability
  • Add to Cart