ENA371178006 Enamine

2-{[(tert-butoxy)carbonyl](2-methylpropyl)amino}acetic acid

  • MDL number MFCD17286086
  • Cas Number : 194206-80-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-107855

Safety Information

msds https://www.enaminestore.com/catalog/EN300-107855
Availability details for
Quantity
  • Availability
  • Add to Cart