ENA371178007 Enamine

{[3-(trifluoromethyl)phenyl]methyl}boranediol

  • MDL number MFCD11856001
  • Cas Number : 21948-55-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 71...73
Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-107857

Safety Information

msds https://www.enaminestore.com/catalog/EN300-107857
Availability details for
Quantity
  • Availability
  • Add to Cart