ENA371178011 Enamine

methyl 4-propanoylbenzoate

  • MDL number MFCD13192729
  • Cas Number : 17745-40-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-105998

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-105998
Availability details for
Quantity
  • Availability
  • Add to Cart