ENA371178013 Enamine

2-(chloromethyl)pyrimidin-4-amine

  • MDL number MFCD10697353
  • Cas Number : 79651-35-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-107884

Safety Information

msds https://www.enaminestore.com/catalog/EN300-107884
Availability details for
Quantity
  • Availability
  • Add to Cart