ENA371178020 Enamine

2-(1-chloroethyl)-5-(3,4-dichlorophenyl)-1,3,4-oxadiazole

  • MDL number MFCD12188339
  • Cas Number : 1156946-42-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 93...95
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-106050

Safety Information

msds https://www.enaminestore.com/catalog/EN300-106050
Availability details for
Quantity
  • Availability
  • Add to Cart