ENA371178021 Enamine

2-bromo-4,6-dimethoxybenzaldehyde

  • MDL number MFCD11110636
  • Cas Number : 81574-69-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-107893

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-107893
Availability details for
Quantity
  • Availability
  • Add to Cart