ENA371178024 Enamine

N1-(cyclohexylmethyl)benzene-1,2-diamine

  • MDL number MFCD12166788
  • Cas Number : 163618-44-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-107897

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-107897
Availability details for
Quantity
  • Availability
  • Add to Cart