ENA371178025 Enamine

3-(3-methylbutyl)piperidine hydrochloride

  • MDL number MFCD22375383
  • Cas Number : 1384669-07-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 174...176
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-107898

Safety Information

msds https://www.enaminestore.com/catalog/EN300-107898
Availability details for
Quantity
  • Availability
  • Add to Cart