ENA371178027 Enamine

4-(2,5-dimethyl-1H-pyrrol-1-yl)cyclohexan-1-amine

  • MDL number MFCD16807716
  • Cas Number : 1248224-99-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-107900

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-107900
Availability details for
Quantity
  • Availability
  • Add to Cart