ENA371178031 Enamine

4-(2-methylpropyl)piperidine-2,6-dione

  • MDL number MFCD18277013
  • Cas Number : 916982-10-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 138-139
Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-107905

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-107905
Availability details for
Quantity
  • Availability
  • Add to Cart