ENA371178039 Enamine

2-cyclobutyl-1,3-thiazole-5-carbaldehyde

  • MDL number MFCD21117859
  • Cas Number : 1394042-39-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-107926

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-107926
Availability details for
Quantity
  • Availability
  • Add to Cart