ENA371178055 Enamine

5-(methoxymethyl)-2,3-dihydro-1,3,4-oxadiazol-2-one

  • MDL number MFCD19557534
  • Cas Number : 1711-90-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-106082

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-106082
Availability details for
Quantity
  • Availability
  • Add to Cart