ENA371178072 Enamine

1-(furan-2-yl)-2-(pyrrolidin-2-yl)ethan-1-one hydrochloride

  • MDL number MFCD22375238
  • Cas Number : 1384676-69-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-106152

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-106152
Availability details for
Quantity
  • Availability
  • Add to Cart