ENA371178080 Enamine

N-[4-(2-chloroacetyl)phenyl]-N-methylformamide

  • MDL number MFCD11104739
  • Cas Number : 1152591-86-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-108074

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-108074
Availability details for
Quantity
  • Availability
  • Add to Cart