ENA371178084 Enamine

2-(chloromethyl)-5-[(2-methylphenyl)methyl]-1,3,4-oxadiazole

  • MDL number MFCD11213536
  • Cas Number : 1094697-61-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-108078

Safety Information

signalword Danger
pcodes P261,P264,P271,P280,P302+P352,P304+P340,P305+P351+P338,P310,P312,P332+P313,P362,P363,P403+P233,P405,P501
hcodes Xi,Xn
MSDS https://www.enaminestore.com/catalog/EN300-108078
pictograms GHS05,GHS07
hstatements H312,H315,H318,H335
Availability details for
Quantity
  • Availability
  • Add to Cart