ENA371178089 Enamine

7-amino-2-methyl-1,2,3,4-tetrahydroisoquinolin-1-one

  • MDL number MFCD19347475
  • Cas Number : 643087-30-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 127...129
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-108084

Safety Information

msds https://www.enaminestore.com/catalog/EN300-108084
Availability details for
Quantity
  • Availability
  • Add to Cart