ENA371178091 Enamine

3-(benzylamino)-2-methylbutan-2-ol

  • MDL number MFCD21106820
  • Cas Number : 63557-73-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-108086

Safety Information

msds https://www.enaminestore.com/catalog/EN300-108086
Availability details for
Quantity
  • Availability
  • Add to Cart