ENA371178101 Enamine

5-[(4-methoxyphenyl)carbamoyl]thiophene-2-carboxylic acid

  • MDL number MFCD12465704
  • Cas Number : 1154324-72-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-108099

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-108099
Availability details for
Quantity
  • Availability
  • Add to Cart