ENA371178104 Enamine

7-[(2-methoxyethyl)amino]-6-nitro-3,4-dihydroquinazolin-4-one

  • MDL number MFCD12739537
  • Cas Number : 1156195-94-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 259...261
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-108116

Safety Information

msds https://www.enaminestore.com/catalog/EN300-108116
Availability details for
Quantity
  • Availability
  • Add to Cart