ENA371178120 Enamine

2-chloro-1-(2-chloro-4-fluoro-5-methylphenyl)ethan-1-one

  • MDL number MFCD22375220
  • Cas Number : 154258-17-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 36...38
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-102190

Safety Information

msds https://www.enaminestore.com/catalog/EN300-102190
Availability details for
Quantity
  • Availability
  • Add to Cart