ENA371178123 Enamine

N,N-dimethyl-2-(N-methylacetamido)acetamide

  • MDL number MFCD20355937
  • Cas Number : 29399-02-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-108157

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-108157
Availability details for
Quantity
  • Availability
  • Add to Cart