ENA371178125 Enamine

methyl 5-amino-2-tert-butyl-1,3-oxazole-4-carboxylate

  • MDL number MFCD20726958
  • Cas Number : 785805-35-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-108159

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-108159
Availability details for
Quantity
  • Availability
  • Add to Cart