ENA371178129 Enamine

(2-nitrophenyl)methanesulfonamide

  • MDL number MFCD12783430
  • Cas Number : 51145-00-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 131...133
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-108163

Safety Information

msds https://www.enaminestore.com/catalog/EN300-108163
Availability details for
Quantity
  • Availability
  • Add to Cart