ENA371178131 Enamine

N,N-dimethyl-3-(piperazin-1-yl)propanamide

  • MDL number MFCD09996861
  • Cas Number : 89009-68-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-108166

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-108166
Availability details for
Quantity
  • Availability
  • Add to Cart