ENA371178137 Enamine

5-(4-fluorophenyl)-1,2,4-oxadiazole-3-carboxylic acid

  • MDL number MFCD00974434
  • Cas Number : 72542-80-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 129...131
Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-108173

Safety Information

msds https://www.enaminestore.com/catalog/EN300-108173
Availability details for
Quantity
  • Availability
  • Add to Cart