ENA371178140 Enamine

1-tert-butyl-4-chloro-6-methyl-1H-pyrazolo[3,4-d]pyrimidine

  • MDL number MFCD11207439
  • Cas Number : 1094418-20-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-108178

Safety Information

msds https://www.enaminestore.com/catalog/EN300-108178
Availability details for
Quantity
  • Availability
  • Add to Cart