ENA371178141 Enamine

2-(3-chlorophenyl)-2-(piperazin-1-yl)ethan-1-ol

  • MDL number MFCD12826665
  • Cas Number : 1183442-35-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-108179

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-108179
Availability details for
Quantity
  • Availability
  • Add to Cart