ENA371178143 Enamine

(3-chlorophenyl)(cyclohexyl)methanamine

  • MDL number MFCD12153888
  • Cas Number : 1183190-35-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-108213

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-108213
Availability details for
Quantity
  • Availability
  • Add to Cart