ENA371178147 Enamine

[1-(2-methoxyethyl)cyclobutyl]methanamine

  • MDL number MFCD21131483
  • Cas Number : 1023888-30-3
  • Molecular Weight : 143.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name [1-(2-methoxyethyl)cyclobutyl]methanamine
Inchi Code 1S/C8H17NO/c1-10-6-5-8(7-9)3-2-4-8/h2-7,9H2,1H3
Inchi Key DUZWPHTXSOEBCA-UHFFFAOYSA-N

Description

Purity 95
Storage temp. +4
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-108217

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-108217
Availability details for
Quantity
  • Availability
  • Add to Cart