ENA371178166 Enamine

1-(6-chloro-1-methyl-1H-1,3-benzodiazol-2-yl)ethan-1-one

  • MDL number MFCD19288835
  • Cas Number : 1145670-42-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-108243

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-108243
Availability details for
Quantity
  • Availability
  • Add to Cart