ENA371178169 Enamine

3-(chloromethyl)-5-(4-methyl-1,3-thiazol-5-yl)-1,2,4-oxadiazole

  • MDL number MFCD10693766
  • Cas Number : 1152504-93-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 82...84
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-108246

Safety Information

msds https://www.enaminestore.com/catalog/EN300-108246
Availability details for
Quantity
  • Availability
  • Add to Cart