ENA371178170 Enamine

5-bromo-2-methoxy-N-(pyridin-4-ylmethyl)aniline

  • MDL number MFCD13474146
  • Cas Number : 1303917-46-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 89...91
Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-106336

Safety Information

msds https://www.enaminestore.com/catalog/EN300-106336
Availability details for
Quantity
  • Availability
  • Add to Cart