ENA371178171 Enamine

2-(3-nitro-1H-pyrazol-1-yl)cyclohexan-1-one

  • MDL number MFCD22375244
  • Cas Number : 1384582-16-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-106344

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-106344
Availability details for
Quantity
  • Availability
  • Add to Cart