ENA371178172 Enamine

ethyl 2-(5,6-dimethyl-1H-1,3-benzodiazol-2-yl)acetate

  • MDL number MFCD22375245
  • Cas Number : 2148-86-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 169...171
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-106347

Safety Information

msds https://www.enaminestore.com/catalog/EN300-106347
Availability details for
Quantity
  • Availability
  • Add to Cart