ENA371178173 Enamine

5-(2,6-dichlorophenoxymethyl)-1,2-oxazole-3-carboxylic acid

  • MDL number MFCD13481123
  • Cas Number : 1225900-23-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 159...161
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-106348

Safety Information

msds https://www.enaminestore.com/catalog/EN300-106348
Availability details for
Quantity
  • Availability
  • Add to Cart