ENA371178176 Enamine

(3-chlorophenyl)(cyclohexyl)methanone

  • MDL number MFCD03841352
  • Cas Number : 211985-77-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-108254

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-108254
Availability details for
Quantity
  • Availability
  • Add to Cart