ENA371178179 Enamine

3-fluoro-4-methanesulfonylbenzene-1-sulfonyl chloride

  • MDL number MFCD21235079
  • Cas Number : 1394041-07-6
  • Molecular Weight : 272.71

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-108307

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-108307
Availability details for
Quantity
  • Availability
  • Add to Cart