ENA371178190 Enamine

1-(furan-2-yl)-2-(pyrrolidin-2-yl)ethan-1-ol

  • Molecular Weight : 181.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1-(2-furyl)-2-(2-pyrrolidinyl)ethanol
Inchi Code 1S/C10H15NO2/c12-9(10-4-2-6-13-10)7-8-3-1-5-11-8/h2,4,6,8-9,11-12H,1,3,5,7H2
Inchi Key WFLJBRRIRXFMNI-UHFFFAOYSA-N

Description

Storage temp. RT
Physical Form oil
Shipping Temperature Normal
Country of Origin UA

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-106370
hstatements H302,H315,H318,H335
Availability details for
Quantity
  • Availability
  • Add to Cart