ENA371178191 Enamine

2-acetamido-N,N-dimethylacetamide

  • MDL number MFCD19159613
  • Cas Number : 29824-30-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-108322

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-108322
Availability details for
Quantity
  • Availability
  • Add to Cart